Gene Transcripts
- Gene Symbol
- NORAD
- Entrez Gene ID
- 647979
- Gene Name
- non-coding RNA activated by DNA damage
- Gene Aliases
- LINC00657
- Chromosome Location
- Chr. 20: 36045618 - 36050960 on Build GRCh38
- UniGene ID
- Hs.743232
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027451.1 |
|
1 |
3311 |
|
View Full Product Details