Gene Transcripts

Gene Symbol
Nct1
Entrez Gene ID
100144395
Gene Name
non-coding in testis 1
Gene Aliases
Chromosome Location
Chr. : - on Build
UniGene ID
Mm.424931
Species
Mus musculus
Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AY940662.1 - LIT3382
Nct1
1 366

View Full Product Details