Gene Transcripts

Gene Symbol
LMF1
Entrez Gene ID
64788
Gene Name
lipase maturation factor 1
Gene Aliases
C16orf26, HMFN1876, JFP11, TMEM112, TMEM112A
Chromosome Location
Chr. 16: 853632 - 981615 on Build GRCh38
UniGene ID
Hs.71912
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_036442.1 4 1601
GenBank mRNA BC036550.1 - 2 3574
BC047239.1 - 3 3187
BC070105.1 - 2 3072
BC098409.1 - 2 3072

View Full Product Details