Gene Transcripts

Gene Symbol
RAB2A
Entrez Gene ID
5862
Gene Name
RAB2A, member RAS oncogene family
Gene Aliases
LHX, RAB2
Chromosome Location
Chr. 8: 60516910 - 60623644 on Build GRCh38
UniGene ID
Hs.369017
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242644.1 NP_001229573.1 7 2205
NM_002865.2 NP_002856.1 8 2277
GenBank mRNA AL137321.1 - 1 1013

View Full Product Details