Gene Transcripts

Gene Symbol
GS1-204I12.4
Entrez Gene ID
105371653
Gene Name
uncharacterized GS1-204I12.4
Gene Aliases
Chromosome Location
Chr. 1: 185646373 - 185657168 on Build GRCh38
UniGene ID
Hs.738511
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF508908.1 - 2 313


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BE906931.1 - 3 340 Chr. 1 - 185651167 - 185656997 [+]

View Full Product Details