Gene Transcripts
- Gene Symbol
- JAKMIP3
- Entrez Gene ID
- 282973
- Gene Name
- Janus kinase and microtubule interacting protein 3
- Gene Aliases
- C10orf14, C10orf39, Jamip3, NECC2, bA140A10.5
- Chromosome Location
- Chr. 10: 132036068 - 132184852 on Build GRCh38
- UniGene ID
- Hs.106254
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK123426.1 |
- |
1 |
2392 |
|
AL832756.1 |
- |
1 |
888 |
|
View Full Product Details