Gene Transcripts

Gene Symbol
ALDH7A1
Entrez Gene ID
501
Gene Name
aldehyde dehydrogenase 7 family member A1
Gene Aliases
ATQ1, EPD, PDE
Chromosome Location
Chr. 5: 126541841 - 126595390 on Build GRCh38
UniGene ID
Hs.483239
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001182.4 NP_001173.2 18 4891
NM_001201377.1 NP_001188306.1 18 4891
NM_001202404.1 NP_001189333.2 16 4699
GenBank mRNA AK123774.1 - 1 1250

View Full Product Details