Gene Transcripts
- Gene Symbol
- METTL3
- Entrez Gene ID
- 56339
- Gene Name
- methyltransferase like 3
- Gene Aliases
- IME4, M6A, MT-A70, Spo8
- Chromosome Location
- Chr. 14: 21498131 - 21511375 on Build GRCh38
- UniGene ID
- Hs.168799
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK097873.1 |
- |
4 |
2118 |
|
BX247964.1 |
- |
8 |
1332 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AK124183.1 |
- |
1 |
1030 |
|
Chr. 14 - 21498139 - 21500234 [-] |
View Full Product Details