Gene Transcripts
- Gene Symbol
- MYCN
- Entrez Gene ID
- 4613
- Gene Name
- v-myc avian myelocytomatosis viral oncogene neuroblastoma derived homolog
- Gene Aliases
- MODED, N-myc, NMYC, ODED, bHLHe37
- Chromosome Location
- Chr. 2: 15940438 - 15947007 on Build GRCh38
- UniGene ID
- Hs.25960
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AA609982.1 |
- |
2 |
192 |
|
Chr. 2 - 15940185 - 15942249 [+] |
View Full Product Details