Gene Transcripts
- Gene Symbol
- BPIFA2
- Entrez Gene ID
- 140683
- Gene Name
- BPI fold containing family A member 2
- Gene Aliases
- C20orf70, PSP, SPLUNC2, bA49G10.1
- Chromosome Location
- Chr. 20: 33161848 - 33181417 on Build GRCh38
- UniGene ID
- Hs.125960
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK130550.1 |
- |
1 |
1402 |
|
View Full Product Details