Gene Transcripts
- Gene Symbol
- FUNDC2
- Entrez Gene ID
- 65991
- Gene Name
- FUN14 domain containing 2
- Gene Aliases
- DC44, HCBP6, HCC3, PD03104
- Chromosome Location
- Chr. X: 155026789 - 155056916 on Build GRCh38
- UniGene ID
- Hs.356050
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF267862.1 |
- |
1 |
12 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AA258633.1 |
- |
1 |
12 |
|
Chr. 5 - 73508509 - 73508886 [+] |
View Full Product Details