Gene Transcripts
- Gene Symbol
- CATX-1
- Entrez Gene ID
- 100506513
- Gene Name
- uncharacterized LOC100506513
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.676466
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF083117.1 |
- |
1 |
375 |
|
View Full Product Details