Gene Transcripts
- Gene Symbol
- DOCK9
- Entrez Gene ID
- 23348
- Gene Name
- dedicator of cytokinesis 9
- Gene Aliases
- ZIZ1, ZIZIMIN1
- Chromosome Location
- Chr. 13: 98793487 - 99086406 on Build GRCh38
- UniGene ID
- Hs.596105
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF339774.1 |
- |
1 |
341 |
|
AK022011.1 |
- |
1 |
1492 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BX100976.1 |
- |
1 |
341 |
|
Chr. 13 - 98849432 - 98850065 [-] |
View Full Product Details