Gene Transcripts
- Gene Symbol
- HSPE1P13
- Entrez Gene ID
- 106480256
- Gene Name
- heat shock protein family E (Hsp10) member 1 pseudogene 13
- Gene Aliases
- Chromosome Location
- Chr. 2: 39098149 - 39098445 on Build GRCh38
- UniGene ID
- Hs.688190
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC047033.1 |
- |
1 |
414 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BG122370.1 |
- |
1 |
408 |
|
Chr. 2 - 39097991 - 39098686 [-] |
View Full Product Details