Gene Transcripts
- Gene Symbol
- SDCCAG8
- Entrez Gene ID
- 10806
- Gene Name
- serologically defined colon cancer antigen 8
- Gene Aliases
- BBS16, CCCAP, CCCAP SLSN7, HSPC085, NPHP10, NY-CO-8, SLSN7, hCCCAP
- Chromosome Location
- Chr. 1: 243255328 - 243500092 on Build GRCh38
- UniGene ID
- Hs.591530
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC015867.1 |
- |
1 |
501 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BX111512.1 |
- |
1 |
441 |
|
Chr. 1 - 243256097 - 243256599 [+] |
View Full Product Details