Gene Transcripts
- Gene Symbol
- CTNNBIP1
- Entrez Gene ID
- 56998
- Gene Name
- catenin beta interacting protein 1
- Gene Aliases
- ICAT
- Chromosome Location
- Chr. 1: 9848276 - 9910322 on Build GRCh38
- UniGene ID
- Hs.463759
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC035794.1 |
- |
2 |
707 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BM557965.1 |
- |
2 |
696 |
|
Chr. 1 - 9880099 - 9910219 [-] |
View Full Product Details