Gene Transcripts
- Gene Symbol
- FTCD
- Entrez Gene ID
- 10841
- Gene Name
- formimidoyltransferase cyclodeaminase
- Gene Aliases
- LCHC1
- Chromosome Location
- Chr. 21: 46135981 - 46156481 on Build GRCh38
- UniGene ID
- Hs.415846
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AL109817.1 |
- |
1 |
219 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
N76219.1 |
- |
2 |
189 |
|
Chr. 21 - 46138527 - 46139271 [-] |
View Full Product Details