Gene Transcripts
- Gene Symbol
- SPATA22
- Entrez Gene ID
- 84690
- Gene Name
- spermatogenesis associated 22
- Gene Aliases
- NYD-SP20, NYDSP20
- Chromosome Location
- Chr. 17: 3440011 - 3513852 on Build GRCh38
- UniGene ID
- Hs.351068
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK057485.1 |
- |
1 |
130 |
|
View Full Product Details