Gene Transcripts

Gene Symbol
NEMF
Entrez Gene ID
9147
Gene Name
nuclear export mediator factor
Gene Aliases
NY-CO-1, SDCCAG1
Chromosome Location
Chr. 14: 49782083 - 49853073 on Build GRCh38
UniGene ID
Hs.655964
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017021760.1 XP_016877249.1 32 5361
XM_017021761.1 XP_016877250.1 27 4814


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AL547508.3 - 1 501 Chr. 14 - 49782094 - 49783001 [-]
GenBankmRNA BQ189493.1 - 1 166 Chr. 14 - 49782083 - 49782666 [-]
GenBankmRNA CR622792.1 - 1 501 Chr. 14 - 49782083 - 49783001 [-]
GenBankmRNA BM455267.1 - 1 180 Chr. 14 - 49781683 - 49782680 [-]

View Full Product Details