Gene Transcripts
- Gene Symbol
- EHF
- Entrez Gene ID
- 26298
- Gene Name
- ETS homologous factor
- Gene Aliases
- ESE3, ESE3B, ESEJ
- Chromosome Location
- Chr. 11: 34621041 - 34663288 on Build GRCh38
- UniGene ID
- Hs.653859
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK025244.1 |
- |
1 |
2476 |
|
View Full Product Details