Gene Transcripts
- Gene Symbol
- FIS1
- Entrez Gene ID
- 51024
- Gene Name
- fission, mitochondrial 1
- Gene Aliases
- CGI-135, TTC11
- Chromosome Location
- Chr. 7: 101239612 - 101245090 on Build GRCh38
- UniGene ID
- Hs.423968
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK310968.1 |
- |
1 |
75 |
|
AK311665.1 |
- |
1 |
56 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AK091729.1 |
- |
1 |
83 |
|
Chr. 7 - 101250335 - 101252320 [-] |
View Full Product Details