Gene Transcripts

Gene Symbol
PSPH
Entrez Gene ID
5723
Gene Name
phosphoserine phosphatase
Gene Aliases
PSP, PSPHD
Chromosome Location
Chr. 7: 56010363 - 56051604 on Build GRCh38
UniGene ID
Hs.512656
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017012467.1 XP_016867956.1 2 585
GenBank mRNA AK054677.1 - 3 657
AK126156.1 - 1 674
BC052958.1 - 3 675

View Full Product Details