Gene Transcripts
- Gene Symbol
- RPL15
- Entrez Gene ID
- 6138
- Gene Name
- ribosomal protein L15
- Gene Aliases
- DBA12, EC45, L15, RPL10, RPLY10, RPYL10
- Chromosome Location
- Chr. 3: 23916804 - 23923696 on Build GRCh38
- UniGene ID
- Hs.381219
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BX537495.1 |
- |
2 |
1067 |
|
View Full Product Details