Gene Transcripts
- Gene Symbol
- POLR2F
- Entrez Gene ID
- 5435
- Gene Name
- RNA polymerase II subunit F
- Gene Aliases
- HRBP14.4, POLRF, RPABC14.4, RPABC2, RPB14.4, RPB6, RPC15
- Chromosome Location
- Chr. 22: 37953663 - 38041915 on Build GRCh38
- UniGene ID
- Hs.436578
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK026327.1 |
- |
1 |
692 |
|
View Full Product Details