Gene Transcripts
- Gene Symbol
- FBXW8
- Entrez Gene ID
- 26259
- Gene Name
- F-box and WD repeat domain containing 8
- Gene Aliases
- FBW6, FBW8, FBX29, FBXO29, FBXW6
- Chromosome Location
- Chr. 12: 116910949 - 117032498 on Build GRCh38
- UniGene ID
- Hs.624537
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC045190.2 |
- |
1 |
2222 |
|
View Full Product Details