Gene Transcripts

Gene Symbol
UGCG
Entrez Gene ID
7357
Gene Name
UDP-glucose ceramide glucosyltransferase
Gene Aliases
GCS, GLCT1
Chromosome Location
Chr. 9: 111896766 - 111935374 on Build GRCh38
UniGene ID
Hs.304249
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003358.2 NP_003349.1 9 2424
XM_005252186.4 XP_005252243.1 10 2447
GenBank mRNA AJ420423.1 - 1 147

View Full Product Details