Gene Transcripts
- Gene Symbol
- CYP19A1
- Entrez Gene ID
- 1588
- Gene Name
- cytochrome P450 family 19 subfamily A member 1
- Gene Aliases
- ARO, ARO1, CPV1, CYAR, CYP19, CYPXIX, P-450AROM
- Chromosome Location
- Chr. 15: 51208057 - 51338598 on Build GRCh38
- UniGene ID
- Hs.260074
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC022896.1 |
- |
3 |
361 |
|
BC056258.1 |
- |
2 |
270 |
|
DA853862.1 |
- |
2 |
265 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
S96437.1 |
- |
1 |
770 |
|
Chr. 15 - 51255415 - 51256271 [-] |
View Full Product Details