Gene Transcripts
- Gene Symbol
- EXTL3-AS1
- Entrez Gene ID
- 101929402
- Gene Name
- EXTL3 antisense RNA 1
- Gene Aliases
- C8orf50
- Chromosome Location
- Chr. 8: 28698403 - 28701464 on Build GRCh38
- UniGene ID
- Hs.667366
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_126027.1 |
|
2 |
666 |
|
GenBank mRNA |
BC118569.1 |
- |
2 |
666 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000520023.1 |
- |
2 |
666 |
|
Chr. 8 - 28698403 - 28701464 [-] |
View Full Product Details