Gene Transcripts
- Gene Symbol
- Clcn5
- Entrez Gene ID
- 12728
- Gene Name
- chloride channel 5
- Gene Aliases
- 5430408K11Rik, ClC-5, Clc5, D930009B12Rik, DXImx42e, Sfc13, T25545
- Chromosome Location
- Chr. X: 7153810 - 7319359 on Build GRCm38
- UniGene ID
- Mm.486257
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK017285.1 |
- |
1 |
107 |
|
View Full Product Details