Gene Transcripts

Gene Symbol
Rapgef6
Entrez Gene ID
192786
Gene Name
Rap guanine nucleotide exchange factor (GEF) 6
Gene Aliases
A530068K01, AI844632, BB085664, C030018K18Rik, PDZ-GEF2, Pdzgef2, RA-GEF-2, Ragef2, mKIAA4052
Chromosome Location
Chr. 11: 54521754 - 54699286 on Build GRCm38
UniGene ID
Mm.254404
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK021094.1 - 1 97
AK038235.1 - 1 706
AK049669.1 - 1 717
AK164343.1 - 1 689

View Full Product Details