Gene Transcripts
- Gene Symbol
- Gabpb2
- Entrez Gene ID
- 213054
- Gene Name
- GA repeat binding protein, beta 2
- Gene Aliases
- 1810015F01Rik, 5830427M07, 9430006E19Rik, A430024B14Rik, AV050852, Gabpb2-1
- Chromosome Location
- Chr. 3: 95181766 - 95217951 on Build GRCm38
- UniGene ID
- Mm.224820
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK007509.1 |
- |
3 |
378 |
|
View Full Product Details