Gene Transcripts

Gene Symbol
Atxn7l3b
Entrez Gene ID
382423
Gene Name
ataxin 7-like 3B
Gene Aliases
4921506J03Rik, 6230409E21Rik, AI315132, ENSMUSG00000074747
Chromosome Location
Chr. 10: 112925428 - 112929026 on Build GRCm38
UniGene ID
Mm.431239
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK037008.1 - 544
AK051537.1 - 1573

View Full Product Details