Gene Transcripts
- Gene Symbol
- Cct4
- Entrez Gene ID
- 12464
- Gene Name
- chaperonin containing Tcp1, subunit 4 (delta)
- Gene Aliases
- 2610204B21Rik, A45, C78323, Cctd
- Chromosome Location
- Chr. 11: 22990593 - 23003336 on Build GRCm38
- UniGene ID
- Mm.296985
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK011873.1 |
- |
|
799 |
|
View Full Product Details