Gene Transcripts
- Gene Symbol
- Acta2
- Entrez Gene ID
- 11475
- Gene Name
- actin, alpha 2, smooth muscle, aorta
- Gene Aliases
- 0610041G09Rik, Actvs, SMalphaA, a-SMA, alphaSMA
- Chromosome Location
- Chr. 19: 34240336 - 34255585 on Build GRCm38
- UniGene ID
- Mm.213025
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK077294.1 |
- |
1 |
307 |
|
AK080187.1 |
- |
1 |
860 |
|
AK137170.1 |
- |
1 |
867 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB222563.2 |
- |
1 |
375 |
|
Chr. 19 - 34240388 - 34241053 [-] |
GenBankmRNA |
BY716908.1 |
- |
|
304 |
|
Chr. - - [-] |
View Full Product Details