Gene Transcripts
- Gene Symbol
- Kif24
- Entrez Gene ID
- 109242
- Gene Name
- kinesin family member 24
- Gene Aliases
- 4933425J19Rik, 9430029L23Rik
- Chromosome Location
- Chr. 4: 41390748 - 41464933 on Build GRCm38
- UniGene ID
- Mm.370288
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK020445.1 |
- |
2 |
1038 |
|
BB091323.1 |
- |
1 |
171 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CA872730.1 |
- |
2 |
322 |
|
Chr. 4 - 41454391 - 41460094 [-] |
View Full Product Details