Gene Transcripts
- Gene Symbol
- Tbck
- Entrez Gene ID
- 271981
- Gene Name
- TBC1 domain containing kinase
- Gene Aliases
- 1700120J03Rik, 9430001M19, A630047E20Rik, C030007I09Rik, Tbckl
- Chromosome Location
- Chr. 3: 132683994 - 132841688 on Build GRCm38
- UniGene ID
- Mm.257864
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK046449.1 |
- |
1 |
1671 |
|
AK081215.1 |
- |
1 |
345 |
|
AW125018.1 |
- |
1 |
295 |
|
View Full Product Details