Gene Transcripts
- Gene Symbol
- Fus
- Entrez Gene ID
- 233908
- Gene Name
- fused in sarcoma
- Gene Aliases
- D430004D17Rik, D930039C12Rik, Fus1, Tls
- Chromosome Location
- Chr. 7: 127966788 - 127982032 on Build GRCm38
- UniGene ID
- Mm.277680
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK038226.1 |
- |
6 |
3873 |
|
AK042244.1 |
- |
1 |
854 |
|
AK084867.1 |
- |
1 |
1586 |
|
View Full Product Details