Gene Transcripts

Gene Symbol
Upf1
Entrez Gene ID
19704
Gene Name
UPF1 regulator of nonsense transcripts homolog (yeast)
Gene Aliases
B430202H16Rik, NORF1, PNORF-1, Rent1, Upflp
Chromosome Location
Chr. 8: 70331522 - 70353999 on Build GRCm38
UniGene ID
Mm.390048
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK046599.1 - 1751


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CJ306300.1 - 1 108 Chr. 8 - 70354416 - 70354822 [+]
GenBankmRNA BB325972.2 - 1 382 Chr. 8 - 70354144 - 70354822 [+]

View Full Product Details