Gene Transcripts
- Gene Symbol
- Mbd2
- Entrez Gene ID
- 17191
- Gene Name
- methyl-CpG binding domain protein 2
- Gene Aliases
- MBD2a
- Chromosome Location
- Chr. 18: 70568270 - 70626131 on Build GRCm38
- UniGene ID
- Mm.322
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF072244.1 |
- |
1 |
60 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DT922271.1 |
- |
|
222 |
|
Chr. - - [-] |
View Full Product Details