Gene Transcripts
- Gene Symbol
- Exosc3
- Entrez Gene ID
- 66362
- Gene Name
- exosome component 3
- Gene Aliases
- 2310005D06Rik, AI593501, Rrp40
- Chromosome Location
- Chr. 4: 45316629 - 45320603 on Build GRCm38
- UniGene ID
- Mm.275788
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK016397.1 |
- |
|
806 |
|
View Full Product Details