Gene Transcripts
- Gene Symbol
- Shc4
- Entrez Gene ID
- 271849
- Gene Name
- SHC (Src homology 2 domain containing) family, member 4
- Gene Aliases
- 6230417E10Rik, 9930029B02Rik, Gm685, RaLP
- Chromosome Location
- Chr. 2: 125627447 - 125724160 on Build GRCm38
- UniGene ID
- Mm.425101
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK148297.1 |
- |
1 |
524 |
|
View Full Product Details