Gene Transcripts
- Gene Symbol
- Vipas39
- Entrez Gene ID
- 104799
- Gene Name
- VPS33B interacting protein, apical-basolateral polarity regulator, spe-39 homolog
- Gene Aliases
- 6720456H09Rik, 9330175H22Rik, AI413782, SPE-39, Spe39, Vipar, hSPE-39
- Chromosome Location
- Chr. 12: 87238875 - 87266378 on Build GRCm38
- UniGene ID
- Mm.208750
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK020129.1 |
- |
1 |
489 |
|
View Full Product Details