Gene Transcripts
- Gene Symbol
- Rwdd4a
- Entrez Gene ID
- 192174
- Gene Name
- RWD domain containing 4A
- Gene Aliases
- BC016198, Gm1942, Rwdd4
- Chromosome Location
- Chr. 8: 47533645 - 47552837 on Build GRCm38
- UniGene ID
- Mm.273081
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK038266.1 |
- |
1 |
2422 |
|
View Full Product Details