Gene Transcripts

Gene Symbol
Kcnq2
Entrez Gene ID
16536
Gene Name
potassium voltage-gated channel, subfamily Q, member 2
Gene Aliases
HNSPC, KQT2, Nmf134
Chromosome Location
Chr. 2: 181075579 - 181135289 on Build GRCm38
UniGene ID
Mm.40615
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK080597.1 - 1 696
BC059899.1 - 1 906
BC096016.1 - 1 906
BU701896.1 - 1 375

View Full Product Details