Gene Transcripts
- Gene Symbol
- IGF2-AS
- Entrez Gene ID
- 51214
- Gene Name
- IGF2 antisense RNA
- Gene Aliases
- IGF2-AS1, IGF2AS, PEG8
- Chromosome Location
- Chr. 11: 2140512 - 2148666 on Build GRCh38
- UniGene ID
- Hs.716962
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000445504.2 |
- |
1 |
137 |
|
Chr. 11 - 2140501 - 2148642 [+] |
GENCODE |
ENST00000381361.4 |
- |
1 |
121 |
|
Chr. 11 - 2140517 - 2148666 [+] |
View Full Product Details