Gene Transcripts
- Gene Symbol
- C10orf95
- Entrez Gene ID
- 79946
- Gene Name
- chromosome 10 open reading frame 95
- Gene Aliases
- Chromosome Location
- Chr. 10: 102449837 - 102451543 on Build GRCh38
- UniGene ID
- Hs.225084
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AW001224.1 |
- |
1 |
106 |
|
Chr. 10 - 102449974 - 102450436 [-] |
GenBankmRNA |
AI857605.1 |
- |
1 |
70 |
|
Chr. 10 - 102449974 - 102450400 [-] |
GenBankmRNA |
AI830105.1 |
- |
1 |
126 |
|
Chr. 10 - 102450000 - 102450456 [-] |
View Full Product Details