Gene Transcripts
- Gene Symbol
- ADORA2A-AS1
- Entrez Gene ID
- 646023
- Gene Name
- ADORA2A antisense RNA 1
- Gene Aliases
- C22orf45
- Chromosome Location
- Chr. 22: 24429206 - 24495074 on Build GRCh38
- UniGene ID
- Hs.632761
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_028483.2 |
|
8 |
1853 |
|
GenBank mRNA |
BC045098.1 |
- |
8 |
1594 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000427813.6 |
- |
8 |
1853 |
|
Chr. 22 - 24438276 - 24495074 [-] |
View Full Product Details