Gene Transcripts
- Gene Symbol
- B3GALT5-AS1
- Entrez Gene ID
- 114041
- Gene Name
- B3GALT5 antisense RNA 1
- Gene Aliases
- C21orf88
- Chromosome Location
- Chr. 21: 39597147 - 39612822 on Build GRCh38
- UniGene ID
- Hs.375120
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000380604.7 |
- |
1 |
313 |
|
Chr. 21 - 39605624 - 39612821 [-] |
GENCODE |
ENST00000329618.6 |
- |
|
1038 |
|
Chr. 21 - 39605599 - 39612441 [-] |
View Full Product Details