Gene Transcripts
- Gene Symbol
- LOC401052
- Entrez Gene ID
- 401052
- Gene Name
- uncharacterized LOC401052
- Gene Aliases
- Chromosome Location
- Chr. 3: 10006418 - 10011095 on Build GRCh38
- UniGene ID
- Hs.662766
- Species
- Homo sapiens
- Gene Symbol
- EMC3
- Entrez Gene ID
- 55831
- Gene Name
- ER membrane protein complex subunit 3
- Gene Aliases
- POB, TMEM111
- Chromosome Location
- Chr. 3: 9962680 - 10011248 on Build GRCh38
- UniGene ID
- Hs.475392
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
HY006276.1 |
- |
2 |
249 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000454232.1 |
- |
2 |
210 |
|
Chr. 3 - 10007462 - 10011209 [-] |
View Full Product Details