Gene Transcripts
- Gene Symbol
- ESR2
- Entrez Gene ID
- 2100
- Gene Name
- estrogen receptor 2
- Gene Aliases
- ER-BETA, ESR-BETA, ESRB, ESTRB, Erb, NR3A2
- Chromosome Location
- Chr. 14: 64226712 - 64338631 on Build GRCh38
- UniGene ID
- Hs.660607
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
KC777384.1 |
- |
1 |
388 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000555861.1 |
- |
3 |
648 |
|
Chr. 14 - 64329431 - 64336569 [-] |
View Full Product Details